5-Hydroxyindol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1H-Indol-5-ol, 5-Indolol, Indol-5-ol, 5-Hydroxy-1H-indol

121270010 121272500 121270050
ACRO121270010EA 37.1 EUR
ACRO121270010 ACRO121272500 ACRO121270050
5-Hydroxyindol 97%
5-Hydroxyindol
Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
Schmelzpunkt: 105…110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005677
CAS-Nummer: 1953-54-4
EINECS: 217-782-6

Bestellen

Spezifikation Testergebnisse

Appearance Beige to brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 105 to 110 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR