5-Hydroxyindol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1H-Indol-5-ol, 5-Indolol, Indol-5-ol, 5-Hydroxy-1H-indol

L00858.03 L00858.06
L00858.03EA 89.5 EUR
L00858.03 L00858.06
5-Hydroxyindol ≥98%
5-Hydroxyindol

Purity: 98+% CAS No.: 1953-54-4 Molecular Formula: C8H7NO Formula Weight: 133.15 Storage Temperature: 2°C to 8°C Melting Point: 106-111° Beilstein Reference: 112349 MDL No.: MFCD00005677

Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
Schmelzpunkt: 106…111 °C
MDL: MFCD00005677
CAS-Nummer: 1953-54-4
EINECS: 217-782-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR