Indol ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1H-Indol
A14427.0B
A14427.18
A14427.30
A14427.0BEA
382
EUR
A14427.0B
A14427.18
A14427.30
Indol ≥99%
Indol
Purity: 99%
CAS No.: 120-72-9
Molecular Formula: C8H7N
Formula Weight: 117.15
Flash Point: 121°(249°F)
Odour: Unpleasant
pH: 6
Melting Point: 50-54°
Boiling Point: 253-254°
Beilstein Reference: 107693
Merck Reference: 14,4963
UN No.: 2811
MDL No.: MFCD00005607
Formel:
C₈H₇N Molecular Weight: 117,15 g/mol Siedepunkt: 253…254 °C Schmelzpunkt: 50…54 °C Dichte: 1,220 Flammpunkt: 121 °C (249 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005607 CAS-Nummer: 120-72-9 EINECS: 204-420-7 UN: 2811 ADR: 6.1,III Merck Index: 14,4963 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...