2-Methylindol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Methyl-1H-indole
A10764.0B
A10764.18
A10764.30
A10764.0BEA
407
EUR
A10764.0B
A10764.18
A10764.30
2-Methylindol ≥98%
2-Methylindol
Purity: 98+%
CAS No.: 95-20-5
Molecular Formula: C9H9N
Formula Weight: 131.18
Flash Point: 141°(285°F)
Melting Point: 56-60°
Boiling Point: 272-273°
Beilstein Reference: 109781
MDL No.: MFCD00005616
Formel:
C₉H₉N Molecular Weight: 131.18 g/mol Siedepunkt: 272…273 °C Schmelzpunkt: 56…60 °C Dichte: 1.07 Flammpunkt: 141 °C (285 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005616 CAS-Nummer: 95-20-5 EINECS: 202-398-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...