3-Methylindol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Methyl-1H-indol, Skatol, β-Methylindol

L03890.06 L03890.14
L03890.06EA 30.5 EUR
L03890.06 L03890.14
3-Methylindol ≥99%
3-Methylindol

Purity: 99% CAS No.: 83-34-1 Molecular Formula: C9H9N Formula Weight: 131.18 Flash Point: 132°(269°F) Odour: Stench Boiling Point: 265-268° Beilstein Reference: 111296 Merck Reference: 14,8560 MDL No.: MFCD00005627

Formel: C₉H₉N
Molecular Weight: 131.18 g/mol
Siedepunkt: 265…268 °C
Schmelzpunkt: 94…97 °C
Dichte: 1.003 g/cm³ (99.6 °C)
Flammpunkt: 132 °C (269 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005627
CAS-Nummer: 83-34-1
EINECS: 201-471-7
Merck Index: 12,08700

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR