7-Ethylindol ≥98%
Lieferant: Thermo Scientific
Synonyme:
7-Ethyl-1H-indole
L14875.09
L14875.18
L14875.09EA
69.8
EUR
L14875.09
L14875.18
7-Ethylindol ≥98%
7-Ethylindol
Purity: 98+%
CAS No.: 22867-74-9
Molecular Formula: C10H11N
Formula Weight: 145.21
Refractive Index: 1.6030
Boiling Point: 230°
MDL No.: MFCD00143514
Formel:
C₁₀H₁₁N Molecular Weight: 145,2 g/mol Siedepunkt: 230 °C Dichte: 1,058 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143514 CAS-Nummer: 22867-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...