2-Phenylindol 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Phenyl-1H-indole
B23674.18
B23674.30
B23674.18EA
28.9
EUR
B23674.18
B23674.30
2-Phenylindol 95%
2-Phenylindol
Purity: 95%
CAS No.: 948-65-2
Molecular Formula: C14H11N
Formula Weight: 193.25
Melting Point: 189-190°
Boiling Point: 250°/10mm
MDL No.: MFCD00005608
Formel:
C₁₄H₁₁N Molecular Weight: 193,25 g/mol Siedepunkt: 250 °C (10 mmHg) Schmelzpunkt: 189…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005608 CAS-Nummer: 948-65-2 EINECS: 213-436-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...