1-Isochinolinol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Isocarbostyril, 1-Hydroxyisoquinoline
M-Clarity™ Qualitätsstufe = MQ100
152102-10G
152102-1G
152102-5G
SIAL152102-10GEA
233
EUR
SIAL152102-10G
SIAL152102-1G
SIAL152102-5G
1-Isochinolinol, Sigma-Aldrich®
1-Isochinolinol
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006899 CAS-Nummer: 491-30-5 EINECS: 207-732-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...