Europium(II)bromid ≥99,99% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

Synonyme: Europium dibromide

44431.03 44431.06 44431.14
44431.03EA 103 EUR
44431.03 44431.06 44431.14
Europium(II)bromid ≥99,99% (Metall-Basis)
Europium(II)bromid

Purity: 99.99% CAS No.: 13780-48-8 Molecular Formula: EuBr2 Formula Weight: 311.77 Physical Form: Powder Sensitivity: Hygroscopic MDL No.: MFCD01076548

Formel: Br₂Eu
Molecular Weight: 311,77 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01076548
CAS-Nummer: 13780-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR