Gadolinium(III)bromid, ultra trocken ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Gadolinium (III) bromide
Unter Argon in Ampullen verpackt
35737.04
35737.09
35737.04EA
59
EUR
35737.04
35737.09
Gadolinium(III)bromid, ultra trocken ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Gadolinium(III)bromid
Purity: 99.9%
CAS No.: 13818-75-2
Molecular Formula: GdBr3
Formula Weight: 396.96
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
MDL No.: MFCD00049741
Formel:
Br₃Gd Molecular Weight: 396,96 g/mol Schmelzpunkt: 770 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049741 CAS-Nummer: 13818-75-2 EINECS: 237-494-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...