Dysprosiumtrichlorid, ultra trocken ≥99,98% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Scientific
|
Achtung
|
Unter Argon in Ampullen verpackt
35692.06
35692.14
35692.06EA
273
EUR
35692.06
35692.14
Dysprosiumtrichlorid, ultra trocken ≥99,98% (REO, Basis der Oxide der Metalle der seltenen Erden)
Dysprosiumtrichlorid
Purity: 99.98%
CAS No.: 10025-74-8
Molecular Formula: DyCl3
Formula Weight: 268.86
Physical Form: Powder, Ampouled under argon
Density: 3.67
Melting Point: 718°
Merck Reference: 14,3482
MDL No.: MFCD00010984
Formel:
Cl₃Dy Molecular Weight: 268,86 g/mol Schmelzpunkt: 718 °C Dichte: 3,67 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010984 CAS-Nummer: 10025-74-8 EINECS: 233-039-9 Merck Index: 14,3482 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...