Europium(III)chlorid, ultra trocken ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Scientific
Achtung
|
Unter Argon in Ampullen verpackt
35738.03
35738.06
35738.14
AB207764.0005
35738.03EA
106
EUR
35738.03
35738.06
35738.14
ABCRAB207764.0005
Europium(III)chlorid, ultra trocken ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Europium(III)chlorid
Purity: 99.99%
CAS No.: 10025-76-0
Molecular Formula: EuCl3
Formula Weight: 258.32
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
Solubility: Soluble in water
Merck Reference: 14,3904
MDL No.: MFCD00010994
Formel:
Cl₃Eu Molecular Weight: 258,32 g/mol Schmelzpunkt: 850 °C Dichte: 4,89 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010994 CAS-Nummer: 10025-76-0 EINECS: 233-040-4 Merck Index: 14,3904 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...