Gadolinium(III)chlorid, ultra trocken ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Fisher Scientific
Achtung


Unter Argon in Ampullen verpackt

35736.06 35736.14
35736.06EA 200 EUR
35736.06 35736.14
Gadolinium(III)chlorid, ultra trocken ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Gadolinium(III)chlorid

Purity: 99.99% CAS No.: 10138-52-0 Molecular Formula: GdCl3 Formula Weight: 263.61 Physical Form: Powder, Ampouled Under Argon Sensitivity: Hygroscopic Appearance: White Melting Point: 609° Merck Reference: 14,4326 MDL No.: MFCD00011024

Formel: Cl₃Gd
Schmelzpunkt: 609 °C
Dichte: 4,52 g/cm³ (0 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011024
CAS-Nummer: 10138-52-0
EINECS: 233-386-6
Merck Index: 14,4326

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR