Gadolinium(III)chlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

439770-5G 439770-25G
SIAL439770-5GEA 227 EUR
SIAL439770-5G SIAL439770-25G
Gadolinium(III)chlorid, Sigma-Aldrich®
Gadolinium(III)chlorid
Formel: Cl₃Gd
Molecular Weight: 263,61 g/mol
Schmelzpunkt: 604…614 °C
Dichte: 4,52 g/cm³ (0 °C)
MDL: MFCD00011024
CAS-Nummer: 10138-52-0
EINECS: 233-386-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR