Gadolinium(III)fluorid, wasserfrei ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Scientific
Achtung

Synonyme: Gadoliniumtrifluorid

13655.14 13655.22
13655.14EA 139 EUR
13655.14 13655.22
Gadolinium(III)fluorid, wasserfrei ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Gadolinium(III)fluorid

Purity: 99.9% CAS No.: 13765-26-9 Molecular Formula: GdF3 Formula Weight: 214.25 Physical Form: Crystalline Density: 7.1 Appearance: White Odour: Odourless Melting Point: 1231° MDL No.: MFCD00016113

Formel: F₃Gd
Molecular Weight: 214,25 g/mol
Schmelzpunkt: 1231 °C
Dichte: 7,1
Lagertemperatur: Raumtemperatur
MDL: MFCD00016113
CAS-Nummer: 13765-26-9
EINECS: 237-369-4
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR