Neodymtribromid Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Scientific
Achtung

14606.14 14606.22
14606.14EA 128 EUR
14606.14 14606.22
Neodymtribromid Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Neodymtribromid Hydrat

Purity: 99.9% CAS No.: 29843-90-1 Molecular Formula: NdBr3.xH2O Formula Weight: 383.97(anhy) Physical Form: Crystalline

Formel: Br₃Nd·H₂O
Molecular Weight: 401,97 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00748137
CAS-Nummer: 29843-90-1
EINECS: 236-897-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR