Terbium(III)chlorid, wasserfrei ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Scientific
Achtung

41678.03 41678.06
41678.03EA 84.5 EUR
41678.03 41678.06
Terbium(III)chlorid, wasserfrei ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Terbium(III)chlorid

Purity: 99.9% CAS No.: 10042-88-3 Molecular Formula: TbCl3 Formula Weight: 265.29 Physical Form: -250 Mesh Powder Sensitivity: Hygroscopic Solubility: Soluble in water Merck Reference: 14,9157 MDL No.: MFCD00011258

Formel: Cl₃Tb
Molecular Weight: 265,28 g/mol
Schmelzpunkt: 588 °C
Dichte: 4,35 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011258
CAS-Nummer: 10042-88-3
EINECS: 233-132-4
Merck Index: 14,9157

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR