Cer(IV)oxid ≥96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cerdioxyd
40581.30
40581.A1
40581.30EA
114
EUR
40581.30
40581.A1
Cer(IV)oxid ≥96%
Cer(IV)oxid
Purity: 96%
CAS No.: 1306-38-3
Molecular Formula: CeO2
Formula Weight: 172.12
Physical Form: Powder
Density: 7.132
Appearance: Pale yellow
Odour: Odourless
Melting Point: 2600°
Solubility: Insoluble in water. Soluble in H2SO4 and HNO3
Merck Reference: 14,1989
MDL No.: MFCD00010933
Polishing and deColourizing glass, in heat-resistant alloy coatings, or infrared filters to prevent reflection, and as a catalyst for organic reactions
Formel:
CeO₂ Molecular Weight: 172.11 g/mol Siedepunkt: 3468 Schmelzpunkt: 2600 °C Dichte: 7.132 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010933 CAS-Nummer: 1306-38-3 EINECS: 215-150-4 Merck Index: 12,02035 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...