Erbium(III)oxid ≥99,8% (Metall-Basis)
Lieferant: Thermo Scientific
Achtung
Synonyme: Erbiumoxid, Dierbiumtrioxid
36372.14 36372.22
36372.14EA 175 EUR
36372.14 36372.22
Erbium(III)oxid ≥99,8% (Metall-Basis)
Erbium(III)oxid
Purity: 99.8% CAS No.: 12061-16-4 Molecular Formula: Er2O3 Formula Weight: 382.56 Physical Form: 3-12mm Sintered Lump Density: 8.64 Melting Point: 2400° Merck Reference: 14,3646 MDL No.: MFCD00010991
For vacuum deposition
Formel: Er₂O₃
Molecular Weight: 382,52 g/mol
Schmelzpunkt: 2400 °C
Dichte: 8,64
Lagertemperatur: Raumtemperatur
MDL: MFCD00010991
CAS-Nummer: 12061-16-4
EINECS: 235-045-7
Merck Index: 14,3646