Erbium(III)oxid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Erbiumoxid, Dierbiumtrioxid
637343-10G
637343-50G
SIAL637343-10GEA
309
EUR
SIAL637343-10G
SIAL637343-50G
Erbium(III)oxid, Sigma-Aldrich®
Erbium(III)oxid
Formel:
Er₂O₃ Molecular Weight: 382,52 g/mol Schmelzpunkt: 2420 °C Dichte: 7,36 g/cm³ (20 °C) |
MDL:
MFCD00010991 CAS-Nummer: 12061-16-4 EINECS: 235-045-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...