Ytterbium(III)oxid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Diytterbiumtrioxid, Ytterbiumoxid
M-Clarity™ Qualitätsstufe = MQ100
246999-10G
246999-50G
SIAL246999-10GEA
96.2
EUR
SIAL246999-10G
SIAL246999-50G
Ytterbium(III)oxid, Sigma-Aldrich®
Ytterbium(III)oxid
Formel:
O₃Yb₂ Molecular Weight: 394,08 g/mol Schmelzpunkt: 2346 °C Dichte: 9,2 g/cm³ (20 °C) |
MDL:
MFCD00011290 CAS-Nummer: 1314-37-0 EINECS: 215-234-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...