1-Naphthylboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid
344690010
344690050
AC34469.1
AC34469.10
344690250
ACRO344690010EA
31.9
EUR
ACRO344690010
ACRO344690050
MAYBAC34469.1
MAYBAC34469.10
ACRO344690250
1-Naphthylboronsäure 97%
1-Naphthylboronsäure
Formel:
C₁₀H₉BO₂ Molecular Weight: 171,99 g/mol Schmelzpunkt: 196…200 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00019722 CAS-Nummer: 13922-41-3 |
Spezifikation Testergebnisse
Appearance | Off-white to pink Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...