1-Naphthylboronsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid

011006-5G 011006-25G
FLUO011006-5GEA 31.1 EUR
FLUO011006-5G FLUO011006-25G
1-Naphthylboronsäure
1-Naphthylboronsäure
Formel: C₁₀H₉BO₂
Molecular Weight: 171.99 g/mol
Schmelzpunkt: 259
MDL: MFCD00019722
CAS-Nummer: 13922-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR