2-Naphthylboronsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid
011007-5G
011007-25G
FLUO011007-5GEA
0
EUR
FLUO011007-5G
FLUO011007-25G
2-Naphthylboronsäure
2-Naphthylboronsäure
Formel:
C₁₀H₉BO₂ Molecular Weight: 171.99 g/mol |
MDL:
MFCD00236051 CAS-Nummer: 32316-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...