1-Brom-2-methoxynaphthalin ≥97%

Lieferant: Thermo Scientific
Achtung

A18926.06 A18926.14 A18926.22
A18926.06EA 88.4 EUR
A18926.06 A18926.14 A18926.22
1-Brom-2-methoxynaphthalin ≥97%
1-Bromo-2-methoxynaphthalene

Purity: 97% CAS No.: 3401-47-6 Molecular Formula: C11H9BrO Formula Weight: 237.10 Melting Point: 79-80° Beilstein Reference: 2045600

Formel: C₁₁H₉BrO
Molecular Weight: 237,1 g/mol
Schmelzpunkt: 79…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00055374
CAS-Nummer: 3401-47-6
EINECS: 222-272-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR