2,3-Diaminonaphthalin 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Naphthalin-2,3-diamin, 2,3-Naphthylendiamin

A12993.03 A12993.06 A12993.14
A12993.03EA 84 EUR
A12993.03 A12993.06 A12993.14
2,3-Diaminonaphthalin 97%
2,3-Diaminonaphthalin

Purity: 97% CAS No.: 771-97-1 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 193-196° Solubility: Slightly soluble in water Beilstein Reference: 2206394 MDL No.: MFCD00004116

Formel: C₁₀H₁₀N₂
Molecular Weight: 158,2 g/mol
Siedepunkt: 343 °C (1013 hPa)
Schmelzpunkt: 193…196 °C
Dichte: 1,096
Lagertemperatur: Raumtemperatur
MDL: MFCD00004116
CAS-Nummer: 771-97-1
EINECS: 212-241-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR