1-Brom-4-methylnaphthalin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

B72604-25G B72604-5G
SIALB72604-25GEA 92 EUR
SIALB72604-25G SIALB72604-5G
1-Brom-4-methylnaphthalin, Sigma-Aldrich®
1-Bromo-4-methylnaphthalene
Formel: C₁₁H₉Br
Molecular Weight: 221,1 g/mol
MDL: MFCD00003872
CAS-Nummer: 6627-78-7
EINECS: 229-599-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR