Acenaphthenchinon 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,2-Naphtho[1,8-c,d]cyclopentandion
102190250
102191000
ACRO102190250EA
64.7
EUR
ACRO102190250
ACRO102191000
Acenaphthenchinon 95%
Acenaphthenchinon
Formel:
C₁₂H₆O₂ Siedepunkt: 351 °C (1013 hPa) Schmelzpunkt: 257…263 °C Dichte: 1,48 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003805 CAS-Nummer: 82-86-0 EINECS: 201-441-3 |
Spezifikation Testergebnisse
Appearance | Yellow to ochre Powder and/or granules |
Infrared spectrum | Conforms |
Melting point | 253 to 263 °C |
HPLC | ≥94.0 % |
Absorption Ratio P-15/P+15 | 1.10 to 1.26 |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...