Acenaphthen ≥97%
Lieferant: Thermo Scientific
|
|
Synonyme:
1,8-Ethylennaphthalin, Naphtho[1,8-a,b]cyclopentan, 1,2-Dihydroacenaphthylen
A14922.0B
A14922.30
A14922.0BEA
229
EUR
A14922.0B
A14922.30
Acenaphthen ≥97%
Acenaphthen
Purity: 97%
CAS No.: 83-32-9
Molecular Formula: C12H10
Formula Weight: 154.21
Flash Point: 135°(275°F)
Melting Point: 92-95°
Boiling Point: 278°
Beilstein Reference: 386081
Merck Reference: 14,28
UN No.: 3077
MDL No.: MFCD00003807
Formel:
C₁₂H₁₀ Molecular Weight: 154,21 g/mol Siedepunkt: 278 °C Schmelzpunkt: 92…95 °C Dichte: 1,15 Flammpunkt: 135 °C (275 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003807 CAS-Nummer: 83-32-9 EINECS: 201-469-6 UN: 3077 ADR: 9,III Merck Index: 14,28 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...