Gold(III)bromid ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Goldtribromid

A19514.03 A19514.06
A19514.03EA 140 EUR
A19514.03 A19514.06
Gold(III)bromid ≥99%
Gold(III)bromid

Purity: 99% CAS No.: 10294-28-7 Molecular Formula: AuBr3 Formula Weight: 436.69 Melting Point: ca 160° dec. UN No.: 3260 MDL No.: MFCD00014171

Formel: AuBr₃
Molecular Weight: 436,68 g/mol
Schmelzpunkt: ∼ 160 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014171
CAS-Nummer: 10294-28-7
EINECS: 233-654-2
UN: 3260
ADR: 8,III
Merck Index: 12,04539

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR