Kupfer(II)bromid ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Cupric bromide, Cuprous bromide
A16335.14
A16335.22
A16335.36
A16335.14EA
29.9
EUR
A16335.14
A16335.22
A16335.36
Kupfer(II)bromid ≥99%
Kupfer(II)bromid
Purity: 99%
CAS No.: 7789-45-9
Molecular Formula: CuBr2
Formula Weight: 223.36
Odour: Odourless
Melting Point: 498°
Boiling Point: 900°
Merck Reference: 14,2629
UN No.: 3260
MDL No.: MFCD00010970
Formel:
Br₂Cu Molecular Weight: 223,35 g/mol Siedepunkt: 900 °C Schmelzpunkt: 498 °C Dichte: 4,710 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010970 CAS-Nummer: 7789-45-9 EINECS: 232-167-2 UN: 3260 ADR: 8,III Merck Index: 14,2629 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...