Gold(I)chlorid, Sigma-Aldrich®
Lieferant: Merck
481130-1G
481130-5G
SIAL481130-1GEA
366
EUR
SIAL481130-1G
SIAL481130-5G
Gold(I)chlorid, Sigma-Aldrich®
Gold(I)chlorid
Formel:
AuCl Molecular Weight: 232,42 g/mol Schmelzpunkt: 170 °C Dichte: 7,57 g/cm³ (20 °C) |
MDL:
MFCD00046175 CAS-Nummer: 10294-29-8 EINECS: 233-655-8 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...