Gold(III)chlorid (≥64,4% Au)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Goldtrichlorid
12163.01
12163.03
12163.06
12163.01EA
93
EUR
12163.01
12163.03
12163.06
Gold(III)chlorid (≥64,4% Au)
Gold(III)chlorid
CAS No.: 13453-07-1
Molecular Formula: AuCl3
Formula Weight: 303.33
Physical Form: Crystalline
Density: 3.90
Sensitivity: Hygroscopic
Appearance: Red
Odour: Odourless
Melting Point: 180(subl)
Merck Reference: 14,4521
UN No.: 3260
MDL No.: MFCD00014172
Formel:
AuCl₃ Molecular Weight: 303,32 g/mol Schmelzpunkt: 180 °C (Subl.) Dichte: 3,90 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014172 CAS-Nummer: 13453-07-1 EINECS: 236-623-1 UN: 3260 ADR: 8,III Merck Index: 14,4521 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...