Kupfer(II)chlorid, ultra trocken ≥99,995% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Kupferdichlorid

Unter Argon in Ampullen verpackt

35673.06 35673.14 35673.22
35673.06EA 126 EUR
35673.06 35673.14 35673.22
Kupfer(II)chlorid, ultra trocken ≥99,995% (Metall-Basis)
Kupfer(II)chlorid

Purity: 99.995% CAS No.: 7447-39-4 Molecular Formula: CuCl2 Formula Weight: 134.45 Physical Form: Powder, Ampouled under argon Density: 3.386 Appearance: Yellow-browm Odour: Odourless pH: 3 Melting Point: 620° Boiling Point: 993° dec. Merck Reference: 14,2633 UN No.: 2802 MDL No.: MFCD00010972

Formel: Cl₂Cu
Molecular Weight: 134,45 g/mol
Siedepunkt: 993 °C (Zers.)
Schmelzpunkt: 620 °C
Dichte: 3,386
Lagertemperatur: Raumtemperatur
MDL: MFCD00010972
CAS-Nummer: 7447-39-4
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 14,2633

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR