Gold(III)oxid ≥99,99% (Metall-Basis) Au ≥ 88.6%, Premion®

Lieferant: Thermo Scientific
Achtung

12161.01 12161.03 12161.06
12161.01EA 88.9 EUR
12161.01 12161.03 12161.06
Gold(III)oxid ≥99,99% (Metall-Basis) Au ≥ 88.6%, Premion®
Gold(III)oxid

Purity: 99.99% CAS No.: 1303-58-8 Molecular Formula: Au2O3 Formula Weight: 441.93 Physical Form: Powder Appearance: Brown Odour: Odourless Melting Point: 150° dec. Solubility: Insoluble in water. Soluble in HCl, HNO3, and in NaCN solution Merck Reference: 14,4524 MDL No.: MFCD00014173

Formel: Au₂O₃
Molecular Weight: 441,93 g/mol
Schmelzpunkt: 150 °C (Zersetzung)
Dichte: 3,6 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014173
CAS-Nummer: 1303-58-8
EINECS: 215-122-1
Merck Index: 14,4524

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR