Silber(II)oxid (≥86,6% Ag) (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

40187.14 40187.06 40187.22
40187.14EA 393 EUR
40187.14 40187.06 40187.22
Silber(II)oxid (≥86,6% Ag) (Metall-Basis)
Silber(II)oxid

Purity: 99.9% CAS No.: 1301-96-8 Molecular Formula: AgO Formula Weight: 123.87 Physical Form: -100 Mesh Powder Density: 7.44 Appearance: Grey to black Odour: Odourless Melting Point: 100° dec. Merck Reference: 14,8522 UN No.: 3085 MDL No.: MFCD00044285

Formel: AgO
Molecular Weight: 123,87 g/mol
Schmelzpunkt: >100 °C (Zersetzung)
Dichte: 7,44
Lagertemperatur: Raumtemperatur
MDL: MFCD00044285
CAS-Nummer: 1301-96-8
EINECS: 215-098-2
UN: 3085
ADR: 5.1,II
Merck Index: 14,8522

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR