Kupfer(I)sulfid ≥99,5% (Metall-Basis)
Lieferant: Thermo Scientific
Synonyme:
Dikupfersulfid
14718.18
14718.30
14718.A1
14718.18EA
81
EUR
14718.18
14718.30
14718.A1
Kupfer(I)sulfid ≥99,5% (Metall-Basis)
Kupfer(I)sulfid
Purity: 99.5%
CAS No.: 22205-45-4
Molecular Formula: Cu2S
Formula Weight: 159.14
Physical Form: -200 Mesh Powder
Density: 5.60
Melting Point: ca 1100°
Solubility: Practically insoluble in water, acetic acid. Partially soluble in NH4OH
Merck Reference: 14,2667
MDL No.: MFCD00016062
Luminous paints, electrodes, solar cells, as catalyst
Formel:
Cu₂S Molecular Weight: 159,16 g/mol Schmelzpunkt: ∼ 1100 °C Dichte: 5,60 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016062 CAS-Nummer: 22205-45-4 EINECS: 244-842-9 Merck Index: 14,2667 |