Silber(I)sulfid ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Disilbersulfid

11416.06 11416.14 11416.22
11416.06EA 53.1 EUR
11416.06 11416.14 11416.22
Silber(I)sulfid ≥99%
Silber(I)sulfid

Purity: 99% CAS No.: 21548-73-2 Molecular Formula: Ag2S Formula Weight: 247.80 Physical Form: Powder Density: 7.317 Appearance: Dark grey to black Odour: Odourless Melting Point: 825° Solubility: Soluble in HNO3, alkali cyanide solutions. Insoluble in water Merck Reference: 14,8530 UN No.: 3077 MDL No.: MFCD00003406

Formel: Ag₂S
Molecular Weight: 247,8 g/mol
Schmelzpunkt: 825 °C
Dichte: 7,317
Lagertemperatur: Raumtemperatur
MDL: MFCD00003406
CAS-Nummer: 21548-73-2
EINECS: 244-438-2
ADR: 9,III
Merck Index: 14,8530

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR