Silber(I)sulfid ≥99,995% (Metall-Basis), Premion®

Lieferant: Thermo Scientific
Achtung

Synonyme: Disilbersulfid

12113.06 12113.14
12113.06EA 201 EUR
12113.06 12113.14
Silber(I)sulfid ≥99,995% (Metall-Basis), Premion®
Silber(I)sulfid

Purity: 99.995% CAS No.: 21548-73-2 Molecular Formula: Ag2S Formula Weight: 247.80 Physical Form: Powder Density: 7.317 Appearance: Dark grey to black Odour: Odourless Melting Point: 825° Merck Reference: 14,8530 UN No.: 3077 MDL No.: MFCD00003406

Formel: Ag₂S
Molecular Weight: 247,8 g/mol
Schmelzpunkt: 825 °C
Dichte: 7,317
Lagertemperatur: Raumtemperatur
MDL: MFCD00003406
CAS-Nummer: 21548-73-2
EINECS: 244-438-2
UN: 3077
ADR: 9,III
Merck Index: 14,8530

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR