Silber(I)sulfid ≥99,999% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

Synonyme: Disilbersulfid

45458.14 45458.06
45458.14EA 571 EUR
45458.14 45458.06
Silber(I)sulfid ≥99,999% (Metall-Basis)
Silber(I)sulfid

Purity: 99.999% CAS No.: 21548-73-2 Molecular Formula: Ag2S Formula Weight: 247.80 Physical Form: 6mm & down pieces Density: 7.317 Appearance: Dark grey to black Odour: Odourless Melting Point: 825° Merck Reference: 14,8530 UN No.: 3077 MDL No.: MFCD00003406

Formel: Ag₂S
Molecular Weight: 247,8 g/mol
Schmelzpunkt: 825 °C
Dichte: 7,317
Lagertemperatur: Raumtemperatur
MDL: MFCD00003406
CAS-Nummer: 21548-73-2
EINECS: 244-438-2
UN: 3077
ADR: 9,III
Merck Index: 14,8530

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR