Silber(I)thiocyanat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
299669-10G
299669-50G
SIAL299669-10GEA
127
EUR
SIAL299669-10G
SIAL299669-50G
Silber(I)thiocyanat, Sigma-Aldrich®
Silber(I)thiocyanat
Formel:
AgSCN Molecular Weight: 165,95 g/mol |
MDL:
MFCD00003416 CAS-Nummer: 1701-93-5 EINECS: 216-934-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...