Cadmium(II)bromid, wasserfrei 98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Cadmium bromide
32960.14 32960.22
32960.14EA 60.7 EUR
32960.14 32960.22
Cadmium(II)bromid, wasserfrei 98%
Cadmium(II)bromid
Cadmium bromide is used in process engraving, lithography and photography.
Purity: 98% CAS No.: 7789-42-6 Molecular Formula: CdBr2 Formula Weight: 272.22 Physical Form: Powder Density: 5.192 Appearance: White to pale yellow Odour: Odourless Melting Point: 567° Boiling Point: 863° Solubility: Freely soluble in water, alcohol. Slightly soluble in acetone and ether. Merck Reference: 14,1615 UN No.: 2570 MDL No.: MFCD00015996
Process engraving, lithography and photography
Freely soluble in water, alcohol. Slightly soluble in acetone and ether.
Formel: Br₂Cd
Molecular Weight: 272.22 g/mol
Siedepunkt: 863 °C
Schmelzpunkt: 567
Dichte: 5.192
Lagertemperatur: Raumtemperatur
MDL: MFCD00015996
CAS-Nummer: 7789-42-6
EINECS: 232-165-1
UN: 2570
ADR: 6.1 III
Merck Index: 14,1615