Cadmium(II)bromid, ultra trocken ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Cadmium bromide
Unter Argon in Ampullen verpackt
35682.06 35682.14
35682.06EA 194 EUR
35682.06 35682.14
Cadmium(II)bromid, ultra trocken ≥99,999% (Metall-Basis)
Cadmium(II)bromid
Purity: 99.999% CAS No.: 7789-42-6 Molecular Formula: CdBr2 Formula Weight: 272.22 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.192 Appearance: White to pale yellow Odour: Odourless Melting Point: 567° Boiling Point: 863° Merck Reference: 14,1615 UN No.: 2570 MDL No.: MFCD00015996
Formel: Br₂Cd
Molecular Weight: 272.22 g/mol
Siedepunkt: 863 °C
Schmelzpunkt: 567
Dichte: 5.192
Lagertemperatur: Raumtemperatur
MDL: MFCD00015996
CAS-Nummer: 7789-42-6
EINECS: 232-165-1
UN: 2570
ADR: 6.1 III
Merck Index: 14,1615