Zinkbromid ≥99,9% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Zink(II)bromid, Zinkdibromid

40413.18 40413.30 40413.A1
40413.18EA 56.5 EUR
40413.18 40413.30 40413.A1
Zinkbromid ≥99,9% (Metall-Basis)
Zinkbromid

Purity: 99.9% CAS No.: 7699-45-8 Molecular Formula: ZnBr2 Formula Weight: 225.19 Physical Form: Crystalline Density: 4.201 Refractive Index: 1.5452 Sensitivity: Hygroscopic Appearance: White to pale cream Melting Point: 394° Boiling Point: 650° Merck Reference: 14,10129 UN No.: 3260 MDL No.: MFCD00011294

Formel: Br₂Zn
Molecular Weight: 225,2 g/mol
Siedepunkt: 650 °C
Schmelzpunkt: 394 °C
Dichte: 4,201
Lagertemperatur: Raumtemperatur
MDL: MFCD00011294
CAS-Nummer: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8,II
Merck Index: 14,10129

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR