Cadmium(II)chlorid, wasserfrei ≥99,0% ACS
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Cadmium chloride
36629.36
36629.22
36629.36EA
352
EUR
36629.36
36629.22
Cadmium(II)chlorid, wasserfrei ≥99,0% ACS
Cadmium(II)chlorid
Purity: 99.0% min
CAS No.: 10108-64-2
Molecular Formula: CdCl2
Formula Weight: 183.32
Physical Form: Powder
Density: 4.047
Melting Point: 568°
Boiling Point: 960°
Solubility: Freely soluble in water. Soluble in acetone. Slightly soluble in methanol. Insoluble in ether
Merck Reference: 14,1617
UN No.: 2570
MDL No.: MFCD00010916
Preparation of cadmium sulfide, as analytical lab reagent, photography, manufacturing of special mirrors, ice-nucleating agent, lubricant, to absorb H2S in sulfide analysis
Formel:
CdCl₂ Molecular Weight: 183,32 g/mol Siedepunkt: 960 °C Schmelzpunkt: 568 °C Dichte: 4,047 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010916 CAS-Nummer: 10108-64-2 EINECS: 233-296-7 UN: 2570 ADR: 6.1,III Merck Index: 14,1617 |