Quecksilber(II)chlorid ≥99,5% ACS
Lieferant: Thermo Scientific
Gefahr
Synonyme: Quecksilberdichlorid, Sublimat
12274.14 12274.22 12274.36
12274.14EA 49.9 EUR
12274.14 12274.22 12274.36
Quecksilber(II)chlorid ≥99,5% ACS
Quecksilber(II)chlorid
Purity: 99.5% min CAS No.: 7487-94-7 Molecular Formula: Cl2Hg Formula Weight: 271.50 Physical Form: Powder Density: 5.4 Refractive Index: 1.859 Appearance: White Odour: Odourless Melting Point: 280-282° Boiling Point: 302° Solubility: Soluble in boiling water (solubility in cold water is increased by HCl or alkali chlorides), alcohol, glycerol, methanol, acetone, and ethyl acetate Merck Reference: 14,5876 UN No.: 1624 MDL No.: MFCD00011041
Formel: HgCl₂
Molecular Weight: 271,5 g/mol
Siedepunkt: 302 °C
Schmelzpunkt: 280…282 °C
Dichte: 5,4
Lagertemperatur: Raumtemperatur
MDL: MFCD00011041
CAS-Nummer: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1,II
Merck Index: 14,5876