Cadmium(II)carbonat ≥97%

Lieferant: Thermo Scientific
Achtung

A15812.0B A15812.18 A15812.30
A15812.0BEA 243 EUR
A15812.0B A15812.18 A15812.30
Cadmium(II)carbonat ≥97%
Cadmium(II)carbonat

Purity: 97% CAS No.: 513-78-0 Molecular Formula: CdCO3 Formula Weight: 172.42 Odour: Odourless Melting Point: 500° dec. UN No.: 2570 MDL No.: MFCD00010918

Formel: CCdO₃
Molecular Weight: 172,42 g/mol
Schmelzpunkt: 500 °C (Zers.)
Dichte: 4,26
Lagertemperatur: Raumtemperatur
MDL: MFCD00010918
CAS-Nummer: 513-78-0
EINECS: 208-168-9
UN: 2570
ADR: 6.1,III
Merck Index: 14,1616

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR