Zinkfluorid ≥99,995% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Zinkdifluorid
12986.06
12986.14
12986.22
12986.06EA
80.8
EUR
12986.06
12986.14
12986.22
Zinkfluorid ≥99,995% (Metall-Basis), Puratronic®
Zinkfluorid
Purity: 99.995%
CAS No.: 7783-49-5
Molecular Formula: F2Zn
Formula Weight: 103.37
Physical Form: Powder
Density: 4.95
Appearance: White
Melting Point: 872°
Boiling Point: 1500°
Merck Reference: 14,10136
MDL No.: MFCD00011298
Formel:
F₂Zn Molecular Weight: 103,39 g/mol Siedepunkt: 1500 °C Schmelzpunkt: 872 °C Dichte: 4,95 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011298 CAS-Nummer: 7783-49-5 EINECS: 232-001-9 UN: 3288 ADR: 6.1,III Merck Index: 14,10136 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...