Zinkiodid, ultra trocken ≥99,995% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Zinkdiiodid, Zink(II)iodid
Unter Argon in Ampullen verpackt
35727.06
35727.14
35727.22
35727.06EA
106
EUR
35727.06
35727.14
35727.22
Zinkiodid, ultra trocken ≥99,995% (Metall-Basis)
Zinkiodid
Purity: 99.995%
CAS No.: 10139-47-6
Molecular Formula: ZnI2
Formula Weight: 319.18
Physical Form: Granular, Ampouled under argon
Density: 4.74
Melting Point: 446°
Boiling Point: 625° dec.
Merck Reference: 14,10140
UN No.: 3260
MDL No.: MFCD00011299
Formel:
I₂Zn Molecular Weight: 319,2 g/mol Siedepunkt: 625 °C (Zers.) Schmelzpunkt: 446 °C Dichte: 4,74 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011299 CAS-Nummer: 10139-47-6 EINECS: 233-396-0 UN: 3260 ADR: 8,II Merck Index: 14,10140 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...