Cadmium(II)oxid ≥99,998% (Metall-Basis), Puratronic®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cadmiumoxid

10664.14 10664.22 10664.36 10664.A3
10664.14EA 128 EUR
10664.14 10664.22 10664.36 10664.A3
Cadmium(II)oxid ≥99,998% (Metall-Basis), Puratronic®
Cadmium(II)oxid

Purity: 99.998% CAS No.: 1306-19-0 Molecular Formula: CdO Formula Weight: 128.40 Physical Form: Powder Density: 8.15 Appearance: Red-brown Odour: Odourless Melting Point: 1430° Merck Reference: 14,1623 UN No.: 2570 MDL No.: MFCD00010921

Formel: CdO
Molecular Weight: 128,41 g/mol
Siedepunkt: 1559 °C (1013 hPa)
Schmelzpunkt: 1430 °C
Dichte: 8,15
Lagertemperatur: Raumtemperatur
MDL: MFCD00010921
CAS-Nummer: 1306-19-0
EINECS: 215-146-2
UN: 2570
ADR: 6.1,III
Merck Index: 14,1623

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR